Print…
BCTC ≥98%
Fornecedor: ENZO LIFE SCIENCES
Sinônimos: N-(4-(tert-Butyl)phenyl)-4-(3-chloropyridin-2-yl)piperazine-1-carboxamide
BMLCA2310010 BMLCA2310050
ENZOBMLCA2310010EA 114 EUR
ENZOBMLCA2310010 ENZOBMLCA2310050
BCTC ≥98%
BCTC
TRPV1 antagonist
BCTC is a potent and selective inhibitor of capsaicin-induced (IC50=35 nM) and acid-induced (IC50=6 nM) VR1 activation. It displays analgesic activity in rat models of inflammatory and neuropathic pain. TRPV1 antagonist.
Fórmula: C₂₀H₂₅ClN₄O
Peso Molecular: 372.897 g/mol
Número MDL : MFCD08690556
Número CAS: 393514-24-4