Print…
Niguldipine hydrochloride ≥98%
Fornecedor: ENZO LIFE SCIENCES
BMLCA2160010 BMLCA2160050
ENZOBMLCA2160010EA 110 EUR
ENZOBMLCA2160010 ENZOBMLCA2160050
Niguldipine hydrochloride ≥98%
Niguldipine hydrochloride
Ca²⁺ channel blocker
A classic L-type Ca²⁺ channel blocker (IC50=75nM) which can also inhibit T-type Ca²⁺ channels (IC50=244nM). It is a novel inhibitor of drug transport by p-glycoprotein (both stereoisomers are equipotent at inhibition of p-glycoprotein).
Número CAS: 119934-51-9