Print…
Cylindrospermopsin ≥95% (by HPLC)
Fornecedor: ENZO LIFE SCIENCES
ALX350149C100 ALX350149M001
ENZOALX350149C100EA 460 EUR
ENZOALX350149C100 ENZOALX350149M001
Cylindrospermopsin ≥95% (by HPLC)
Cylindrospermopsin
Protein synthesis inhibitor
Tricyclic alkaloid hepatotoxin. Exhibits a completely different mechanism of toxicity than microcystins. Protein synthesis inhibitor. Might be carcinogenic. Inhibits pyrimidine nucleotide synthesis.
Isolated from Cylindrospermopsis raciborskii.
Número CAS: 143545-90-8