Print…
Patulin ≥98% (por HPLC)
Fornecedor: ENZO LIFE SCIENCES
ALX270111M001 ALX270111M005 ALX270111M025
ENZOALX270111M001EA 51.7 EUR
ENZOALX270111M001 ENZOALX270111M005 ENZOALX270111M025
Patulin ≥98% (por HPLC)
Patulin
Inhibitor of protein farnesylation in a cell free assay. Inhibits incorporation of tritiated mevalonate into proteins in whole cells. Mycotoxin with anti-bacterial, potassium uptake inhibitory and possibly carcinogenic activities. Known contaminant of spoiled apples.
Isolated from Penicillium expansum.
Fórmula: C₇H₆O₄
Peso Molecular: 154.121 g/mol
Número MDL : MFCD00005858
Número CAS: 149-29-1