Print…

Lavendustin A ≥98%

Fornecedor: ENZO LIFE SCIENCES

BMLEI1850001 BMLEI1850005
ENZOBMLEI1850001EA 165 EUR
ENZOBMLEI1850001 ENZOBMLEI1850005
Lavendustin A ≥98%
Lavendustin A

EGFR inhibitor


Cell permeable tyrosine kinase inhibitor with little effect on cAMP-dependent protein kinase (PKA) or protein kinase C (PKC). Potent and selective inhibitor of the EGF receptor tyrosine kinase (IC50=11nM). Binds to a site on the kinase which is distinct from the ATP and peptide substrate binding sites. Inhibits EGF-induced proliferation of cultured human myometrial smooth muscle cells and suppresses VEGF-induced angiogenesis in rats. Effective at 10µM in cultured leech neurons. Also inhibits NMDA-stimulated cGMP production.

Fórmula: C₂₁H₁₉NO₆
Número MDL : MFCD00153822
Número CAS: 125697-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20c49bbc936c0a5b6ed966566c36398ff75fadfd&vid=40385a6621663d519049056dd7b74cce2fd5c254&prid=web&clid=SAPDAM

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR