Print…

HET-0016 ≥98%

Fornecedor: ENZO LIFE SCIENCES

BMLEI3500005 BMLEI3500025
ENZOBMLEI3500005EA 110 EUR
ENZOBMLEI3500005 ENZOBMLEI3500025
HET-0016 ≥98%
HET-0016

Cytochrome P450 inhibitor


HET-0016 is a potent and selective inhibitor of the biosynthesis of 20-HETE acting via inhibition of CYP4A. It inhibits cerebrovascular constriction induced by Ca2+ or arachidonate and inhibits angiogenesis and endothelial cell sprouting in various models.

Fórmula: C₁₂H₁₈N₂O
Número MDL : MFCD00277704
Número CAS: 339068-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=950fac892995ba3945ed94b0d3667cb0bb835b85&vid=7e8f965c549f5b16ea006e39ff0d794d545f04e0&prid=web&clid=SAPDAM

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR