Print…
Dexamethasone ≥99% (por TLC)
Fornecedor: ENZO LIFE SCIENCES
Sinônimos: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
BMLEI1260001 BMLEI1260005
ENZOBMLEI1260001EA 89.1 EUR
ENZOBMLEI1260001 ENZOBMLEI1260005
Dexamethasone ≥99% (por TLC)
Dexamethasone
Glucocorticoid receptor ligand
Anti-inflammatory glucocorticoid with anti-inflammatory activity. Induces the production of phospholipase A2 inhibitory protein (lipocortin). Inhibits induction of nitric oxide synthase (NOS) (IC50=5nM). Induces apoptosis in eosinophils, but inhibits it in neutrophils. Both effects are blocked by RU-486. Modulates osteogenesis of mesenchymal stem cells. Inhibits constitutive OPG mRNA steady-state levels in a dose- and time-dependent fashion, promoting osteoclast formation. Oral dexamethasone administration induces an increase in insulin resistance in humans. Activates Indoleamine 2,3-dioxygenase (IDO) in plasmacytoid dendritic cells in vivo and thus promotes tryptophan catabolism and thereby suppression of T cells.
Fórmula: C₂₂H₂₉FO₅
Peso Molecular: 392.47 g/mol
Número MDL : MFCD00064136
Número CAS: 50-02-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2df05c1caf851acea3505319027eadc29a17a510&vid=2908861702a2afaca0b1f9e1e1e89ed845f628b1&prid=web&clid=SAPDAM