Print…
Cyclosporin H
Fornecedor: ENZO LIFE SCIENCES
ALX380286M001 ALX380286M005
ENZOALX380286M001EA 173 EUR
ENZOALX380286M001 ENZOALX380286M005
Cyclosporin H
Proteínas e Peptídeos
TPA/PMA inhibitor
Immunologically inactive. Does not bind to immunophilin. Potent inhibitor of tumor promoting phorbol ester TPA/PMA (Prod. No. BML-PE160) in mouse skin in vivo. Potent inhibitor of Ca²⁺/calmodulin dependent EF-2 phosphorylation in vitro. Potent and selective antagonist of formyl peptide receptor. Inhibits formyl peptide-induced superoxide formation.
Isolated from Fusarium solani.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e39f9178fdabfc67f826760de2632e95a07e75ab&vid=1d2e43600f5a279f88fc5c3e5d0e39044275d827&prid=web&clid=SAPDAM