Print…
SUMO-1 (93-97)-AMCA
Fornecedor: Enzo Life Sciences
ENZOBMLUW05001000EA 159.5 EUR
ENZOBMLUW05001000 ALEXBMLUW05001000
SUMO-1 (93-97)-AMCA
Proteínas e Peptídeos
SUMO1-AMCA is a synthetic AMCA-peptide substrate based on the C-terminal residues of SUMO1 and is a useful fluorogenic substrate for deSUMOylating enzymes, including SENP1 (BML-UW9760).
  • Provides an alternative substrate to SUMO1-AMC
It has particular utility for the study of deSUMOylating activity where continuous monitoring of activity is essential.