Ac-YVAD-pNA
Fornecedor: ENZO LIFE SCIENCES
ALX260026M001
ALX260026M005
ENZOALX260026M001EA
68.2
EUR
ENZOALX260026M001
ENZOALX260026M005
Ac-YVAD-pNA
Proteínas e Peptídeos
Caspase-1 substrate
Sequence is a modification of pro-interleukin-1beta Asp-116 cleavage site for caspase-1. Similar to Ac-YVAD-AMC but cleavage is monitored colorimetrically at 405 nm.