Print…

Anti-AHR Rabbit Polyclonal Antibody

Fornecedor: ENZO LIFE SCIENCES
BMLSA5500025 BMLSA5500100
ENZOBMLSA5500025EA 160 EUR
ENZOBMLSA5500025 ENZOBMLSA5500100
Anti-AHR Rabbit Polyclonal Antibody
Anticorpos
Host: Rabbit

  • For Immunoprecipitation, Western Blot

Recognizes rat and mouse AHR. Detects a band of ~95kDa by Western blot.

Type: Primary
Antigen: AHR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR