Print…
Anti-AHR Rabbit Polyclonal Antibody
Fornecedor: ENZO LIFE SCIENCES
BMLSA2100025 BMLSA2100100
ENZOBMLSA2100025EA 172 EUR
ENZOBMLSA2100025 ENZOBMLSA2100100
Anti-AHR Rabbit Polyclonal Antibody
Anticorpos
Host: Rabbit
  • For Immunoprecipitation, Western Blot, ChIP
Recognizes mouse AhR. Detects a band of ~95 kDa by Western blot. Does not cross-react with Arnt.
Type: Primary
Antigen: AHR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse