Print…
AMPIGENE® HS Taq Mix
Fornecedor: ENZO LIFE SCIENCES
ENZNUC1010040 ENZNUC1010200 ENZNUC1011000
ENZOENZNUC1010040EA 63.8 EUR
ENZOENZNUC1010040 ENZOENZNUC1010200 ENZOENZNUC1011000
AMPIGENE® HS Taq Mix
Reagentes de ácidos nucleicos End-point PCR Enzymes and Kits
Increased sensitivity standard PCR mix for a broader range of samples, with enhanced speed, yield, and specificity.
AMPIGENE® HS Taq Mix uses the latest developments in polymerase technology and buffer chemistry to optimise and increase PCR sensitivity. The AMPIGENE® HS Taq Mix is a robust mix using hot-start technology to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine. An optimized buffer system allows efficient amplification under fast and standard cycling conditions. The AMPIGENE® HS Taq Mix is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine.
Informações de encomenda: Contains 2X AMPIGENE® HS Taq Mix (AMPIGENE® HS Taq Polymerase, 6 mM MgCl2, 2 mM dNTPs, enhancer, and stabilisers).