Print…
SAVIEW® PLUS HRP Reagent
Fornecedor: ENZO LIFE SCIENCES
ENZACC1020150
ENZOENZACC1020150EA 217 EUR
ENZOENZACC1020150
SAVIEW® PLUS HRP Reagent
Reagentes de ácidos nucleicos Nucleic Acid Modification and Labelling
High sensitivity, low background streptavidin-based nanopolymer detection reagent for use with HIGHDEF® chromogens in ISH and IHC applications.
  • Ready-to-use reagent
  • Streptavidin-based detection with reduced carbohydrate and biotin background
  • Suitable for both manual and automated platforms
  • Suitable for custom multiplex IHC protocols
  • The Streptavidin blocker/diluent further reduces background (sold separately)
  • Customise with HIGHDEF® chromogens of your choice (sold separately)
SAVIEW® PLUS HRP reagent is a ready-to-use streptavidin-based nanopolymer detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the SAVIEW® PLUS HRP reagent can be used for in situ hybridisation detection of biotinylated DNA/RNA probes.
Fornecimento: 150 tests × 150 µl (22,5 ml total volume).