Print…
NOR-3 97 HPLC_ASSAY_METHOD
Fornecedor: ENZO LIFE SCIENCES
ALX430011M005 ALX430011M010
ENZOALX430011M005EA 117 EUR
ENZOALX430011M005 ENZOALX430011M010
NOR-3 97 HPLC_ASSAY_METHOD
NOR-3
Nitric oxide donor
Non-thiol-based nitric oxide (NO) donor. NOR-3 shows a variety of biological and pharmacological effects. Strong vasodilatory effects in rat aorta, rabbit aorta and dog coronary artery; potent inhibitory activities against antiplatelet aggregation and thrombus formation; effective aggregation inhibition of human platelet raising by ADP; cardioprotective effects in the ischemia/reperfusion system; antianginal effect.
Número CAS: 163180-49-2