Print…

HAT activator, CTPB 97 1H-NMR

Fornecedor: ENZO LIFE SCIENCES

Sinônimos: N-(4-Chloro-3-(trifluoromethyl)phenyl)-2-ethoxy-6-pentadecylbenzamide, CTPB

ALX420033M001 ALX420033M005
ENZOALX420033M001EA 73.7 EUR
ENZOALX420033M001 ENZOALX420033M005
HAT activator, CTPB 97 1H-NMR
HAT activator, CTPB

p300 HAT activator.


Potent activator of p300 HAT (histone acetyltransferase), but not of PCAF (p300/CBP-associated factor) HAT activities.


Soluble in DMSO.

Fórmula: C₃₁H₄₃ClF₃NO₂
Peso Molecular: 554.135 g/mol
Temperatura de Armazenamento: Congelador
Número MDL : MFCD08702707
Número CAS: 586976-24-1

Encomendar

Especificações dos Resultados do Teste

Purity ≥97% (¹H-NMR)
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR