Print…
Human recombinant NMNAT3 (from E. coli)
Fornecedor: ENZO LIFE SCIENCES
ALX201238C100
ENZOALX201238C100EA 726 EUR
ENZOALX201238C100
Human recombinant NMNAT3 (from E. coli)
Proteínas e Peptídeos
Produced in E. coli. Full length human NMNAT3 is fused at the N-terminus to a His-tag.