Print…
IL-2 (human), ELISA kit
Fornecedor: ENZO LIFE SCIENCES
ADI900118A
ENZOADI900118AEA 621 EUR
ENZOADI900118A
IL-2 (human), ELISA kit
Ensaios ELISAs
The IL-2 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.
  • Low dynamic range provides sensitive measurements
  • Validated in a wide variety of matrices
  • Ready to use reagents and pre-coated plates reduce errors
For the quantitative determination of human IL-2 in culture supernatants, plasma, and serum.
Fornecimento: Supplied with microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.