Print…
VitraPOR® Filter Candles, Cylindrical Shape
Fornecedor: ROBU GLASFILTER
18403 18404 18402 18340 18341 18342 18343 18344 18400 18401
ROBU18403EA 179 EUR
ROBU18403 ROBU18404 ROBU18402 ROBU18340 ROBU18341 ROBU18342 ROBU18343 ROBU18344 ROBU18400 ROBU18401
VitraPOR® Filter Candles, Cylindrical Shape
Filtros Tubos de Filtração
VitraPOR® cylindrical filter candle-blanks are ideal for custom filter assemblies. The conical shape ensures secure fitting, while the porous glass delivers precise and reliable filtration performance.
  • VitraPOR® porous glass filter
  • Cylindrical shape for tight fit
  • Customisable for various systems
  • High chemical and thermal resistance
  • Suitable for gas and liquid filtration
  • Reusable and autoclavable
  • Uniform pore structure
VitraPOR® filter candle-blanks in cylindrical shape are designed for flexible integration into filtration setups where a press-fit or sealed connection is required. Made of borosilicate glass 3.3 with a homogeneous structure, they ensure reliable filtration of gases and liquids with excellent chemical and thermal durability. These blanks serve as a robust base for custom filter solutions and can be regenerated by thermal treatment or cleaning. Ideal for laboratory, pilot, or production applications requiring compact and efficient filtration elements.
Certificações:
In compliance with ISO 4796-80 and ASTM E128-99 standards.