Print…
Axygen® RIGIPLATE® PCR Microplates for Automation
Fornecedor: Corning
PCR-96-BRF-C-C PCR-96-EPF-B-C PCR-384-FS-C-C PCR-96-EPS-C-C PCR-96-BRF-B-C PCR-384-FS-B-C PCR-96-EPF-C-C PCR-96-EPS-B-C
AXYGPCR-96-BRF-C-CEA 285 EUR
AXYGPCR-96-BRF-C-C AXYGPCR-96-EPF-B-C AXYGPCR-384-FS-C-C AXYGPCR-96-EPS-C-C AXYGPCR-96-BRF-B-C AXYGPCR-384-FS-B-C AXYGPCR-96-EPF-C-C AXYGPCR-96-EPS-B-C
Axygen® RIGIPLATE® PCR Microplates for Automation
Microplacas Placas de PCR
These PP/PC PCR plates have maximum volumes that range from 40 to 330 µl and are compatible with most thermal cyclers.
  • ANSI/SBS foot print
  • Smooth, thin, uniform well walls ensure accurate thermal transfer
  • Wells are slightly raised to accommodate sealing mats, films or foils
  • Plates are skirted to allow barcoding, and include a frosted labelling area
  • Lot tested and certified free from DNase, RNase and human genomic DNA
  • Printed alphanumeric labelling simplifies plate and sample identification
Ideal for high throughput screening, sample preparation and NGA library preparation.
Barcoded: No
Certificações:
DNase/RNAse-free, Human DNA free. ISO 13485 and ISO 8 clean room facility.
Embalagem:
10 plate/pack; 50 plates/case.