Print…
GoTaq® Legionella qPCR and Viability qPCR Kits
Fornecedor: Promega
AM2202 AM2206
PROMAM2202EA 1990 EUR
PROMAM2202 PROMAM2206
GoTaq® Legionella qPCR and Viability qPCR Kits
Reagentes de ácidos nucleicos qPCR/RT-qPCR Enzymes and Kits
Reliable detection of Legionella pneumophila in water supplies is important to prevent occurrence of Legionnaires’ disease.
  • Get same-day results for quicker response and intervention
  • Selectively amplify DNA from live cells, if desired, to evaluate the effectiveness of disinfection and treatment processes
  • Identify L. pneumophila or Legionella spp., L. pneumophila and L. pneumophila Serogroup 1 in a single reaction for efficiency
  • Ships at ambient temperature
Legionella testing is commonly performed using culture-based approaches, which often take 7 to 10 days. PCR-based detection is much faster, but it cannot differentiate between live and dead cells, which can possibly lead to false-positive results. The Legionella qPCR Kits combine the rapid protocols of PCR with the option to discriminate between live and dead cells for fast, sensitive and specific Legionella pneumophila detection. We also offer a multiplex kit that can detect Legionella spp., Legionella pneumophila and Legionella pneumophila serogroup 1 in a single assay.
The GoTaq® Legionella qPCR kits have undergone independent evaluation to validate their performance in detecting Legionella species in water samples. This third-party study, conducted in accordance with ISO 12869:2019 standards, assessed assay specificity and DNA recovery. Key findings demonstrate high specificity, robust assay efficiencies, and consistent DNA recovery.