Print…
HexArmor® Safety Gloves, Helix® 2076, 60660
Fornecedor: UVEX
6066005 6066006 6066007 6066008 6066009 6066010 6066011 6066012 6066013
UVEX6066005EA 0 EUR
UVEX6066005 UVEX6066006 UVEX6066007 UVEX6066008 UVEX6066009 UVEX6066010 UVEX6066011 UVEX6066012 UVEX6066013
HexArmor® Safety Gloves, Helix® 2076, 60660
Luvas
Helix® 2076 gloves feature a 13-gauge HPPE, steel, and fibreglass blend shell with seamless construction that gives wearers' hands advanced cut protection, comfort, and breathability on the job, plus a reinforced thumb saddle patch and a flexible PU palm coating.
  • HPPE and steel fiberglass blend shell offering 360° cut protection
  • Flexible polyurethane palm coating
  • Reinforced thumb crotch area for increased durability
  • Seamless construction delivers high comfort and dexterity
The 13-gauge HPPE and steel blend shell offers 360° cut protection and excellent sensitivity in cut protection level F. Flexible polyurethane palm coating provides superior grip and abrasion resistance. A knit wrist helps prevent dirt and debris from entering the glove. Seamless construction allows enhanced comfort while maintaining dexterity and tactility.