Print…
HexArmor® Protective Sleeves, 60985 Series
Fornecedor: UVEX
6098599 6098500 6098501 6098502 6098503 6098504 6098505
UVEX6098599EA 0 EUR
UVEX6098599 UVEX6098500 UVEX6098501 UVEX6098502 UVEX6098503 UVEX6098504 UVEX6098505
HexArmor® Protective Sleeves, 60985 Series
Luvas
These 9" protective arm guards deliver industrial puncture protection against snags, punctures, abrasions, and cuts over the entire forearm, complete with a spandex wrist insert and thumbhole.
  • SuperFabric® material protects against hazards
  • Innovative industrial puncture protection from wires, metal, wood, and glass
  • Lightweight design is cooler than knit or leather alternatives
  • Launderable
Offering excellent durability in cut protection level F, these sleeves feature one layer of SuperFabric® material to protect against hazards through a configuration of tiny guard plates layered onto performance fabrics. Highly durable and long-lasting protection, with a Spandex wrist insert and thumbhole to keep the sleeve in place.