Print…
Assembly Gloves, phynomic lite w 60041
Fornecedor: UVEX
6004105 6004112
UVEX6004105EA 61.2 EUR
UVEX6004105 UVEX6004112
Assembly Gloves, phynomic lite w 60041
Luvas
The uvex phynomic series is ideal for all types of work where a high level of feeling is required.
  • Aqua-polymer impregnation on palm and fingertips
  • Very high level of breathability with porous coating
  • Outstanding tactile sensation when handling small parts
  • Perfect fit with 3D Ergo technology
These safety gloves are like a 'second skin' and are especially light and flexible, without making hands immediately sweaty. The uvex phynomic lite is extremely lightweight, with a composition that makes it suited to particularly delicate tasks.