Print…
D500 foam Safety Gloves, 60604 Series
Fornecedor: UVEX
6060407 6060408 6060409 6060410 6060411
UVEX6060407EA 271 EUR
UVEX6060407 UVEX6060408 UVEX6060409 UVEX6060410 UVEX6060411
D500 foam Safety Gloves, 60604 Series
Luvas
Made with the latest generation of patented uvex Bamboo TwinFlex® technology, the D500 foam glove combines comfort features such as maximum flexibility, perfect fit, and dexterity with outstanding cut protection performance.
  • Very robust and flexible
  • Excellent blade cut resistance (level D)
  • Very high abrasion resistance
  • Free from pollutants
This glove is highly resistant to abrasion, for prolonged product life and exceptional cost efficiency. A very flexible and sensitive cut protection glove for mechanical activities, featuring high-performance elastomer (HPE) and SoftGrip foam coating on palm and fingertips. Very good grip in dry and slightly damp areas.