Print…
HexArmor® Helix® 2065 Safety Gloves, 60659 Series
Fornecedor: UVEX
60659/6 60659/7 60659/8 60659/9
UVEX60659/6EA 17.8 EUR
UVEX60659/6 UVEX60659/7 UVEX60659/8 UVEX60659/9
HexArmor® Helix® 2065 Safety Gloves, 60659 Series
Luvas
The innovative Helix® 2065 safety glove features an HPPE, fibreglass, and nylon blend shell that provides a seamless construction for enhanced comfort and breathability while a double-dipped flexible sandy nitrile palm coating provides 360° cut protection and water resistance for wearers.
  • HPPE, steel, and nylon blend shell offering 360° cut protection
  • Full flat nitrile coating offers of 360° water resistance
  • Seamless construction delivers high comfort and dexterity
The 13-gauge HPPE, steel, and nylon blend shell provies enhanced comfort and worker compliance. Full flat nitrile coating offers of 360° water resistance. Flexible sandy nitrile palm coating provides superior grip and abrasion resistance in wet conditions. Knit wrist helps prevent dirt and debris from entering the glove.