Print…
Safety Gloves unilite 7710F, 60278
Fornecedor: UVEX
6027810 6027807 6027808 6027809 6027811
UVEX6027810EA 56.6 EUR
UVEX6027810 UVEX6027807 UVEX6027808 UVEX6027809 UVEX6027811
Safety Gloves unilite 7710F, 60278
Luvas
The structure of the unilite 7710F seamless polyester assembly gloves, in combination with their full surface NBR coating with grip finish on the palms, makes them ideal for working in dry, dirty, wet, or oily environments.
  • Seam-free multi-purpose gloves
  • Polyester lining material
  • Very good dexterity up to the fingertips
  • Extremely flexible
Fully coated with nitrile rubber (NBR) for protection against moisture and water reaching the skin, these gloves are ideal for precision mechanical work. Additional sandy nitrile coating on the palms of the hands provides grip and abrasion resistance.