Print…

Anti-CHL1 Rabbit Monoclonal Antibody

Fornecedor: Abcam
AB321870-20UL AB321870-100UL AB321870-1ML
ABCAAB321870-20ULEA 244 EUR
ABCAAB321870-20UL ABCAAB321870-100UL ABCAAB321870-1ML
Anti-CHL1 Rabbit Monoclonal Antibody
Anticorpos
Rabbit monoclonal [EPR29395-579] to CHL1.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CHL1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR