Print…

Anti-HCoV-229E Nucleocapsid protein Human Monoclonal Antibody [clone: abd210]

Fornecedor: Abcam
AB316286-1MG AB316286-100UG
ABCAAB316286-1MGEA 9160 EUR
ABCAAB316286-1MG ABCAAB316286-100UG
Anti-HCoV-229E Nucleocapsid protein Human Monoclonal Antibody [clone: abd210]
Anticorpos
Human monoclonal [abd210] to HCoV-229E Nucleocapsid protein - BSA and Azide free.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HCoV-229E Nucleocapsid protein
Clonality: Monoclonal
Clone: abd210
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR