Print…

Anti-B MyB Rabbit Monoclonal Antibody [clone: EPR26321-89]

Fornecedor: Abcam
AB314863-1MG AB314863-100UG
ABCAAB314863-1MGEA 10120 EUR
ABCAAB314863-1MG ABCAAB314863-100UG
Anti-B MyB Rabbit Monoclonal Antibody [clone: EPR26321-89]
Anticorpos
Rabbit monoclonal [EPR26321-89] to B MyB - BSA and Azide free.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: B MyB
Clonality: Monoclonal
Clone: EPR26321-89
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR