Print…
Anti-CISD1/MitoNEET Rabbit Monoclonal Antibody [clone: EPR29116-27]
Fornecedor: Abcam
AB318199-20UL AB318199-100UL AB318199-1ML
ABCAAB318199-20ULEA 244 EUR
ABCAAB318199-20UL ABCAAB318199-100UL ABCAAB318199-1ML
Anti-CISD1/MitoNEET Rabbit Monoclonal Antibody [clone: EPR29116-27]
Anticorpos
Rabbit monoclonal [EPR29116-27] to CISD1/MitoNEET.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CISD1/MitoNEET
Clonality: Monoclonal
Clone: EPR29116-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat