Print…

Anti-Plasma Kallikrein 1B Rabbit Recombinant Multiclonal Antibody [clone: RM2034]

Fornecedor: Abcam
AB315397-1MG AB315397-100UG
ABCAAB315397-1MGEA 9160 EUR
ABCAAB315397-1MG ABCAAB315397-100UG
Anti-Plasma Kallikrein 1B Rabbit Recombinant Multiclonal Antibody [clone: RM2034]
Anticorpos
Rabbit recombinant multiclonal [RM2034] to Plasma Kallikrein 1B - BSA and Azide free.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Plasma Kallikrein 1B
Clonality: Recombinant Multiclonal
Clone: RM2034
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Mouse;Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR