Print…

Anti-SARS-CoV2 nsp12 Rabbit Monoclonal Antibody [clone: EPR24837-213]

Fornecedor: Abcam
AB315964-20UL AB315964-100UL AB315964-1ML
ABCAAB315964-20ULEA 244 EUR
ABCAAB315964-20UL ABCAAB315964-100UL ABCAAB315964-1ML
Anti-SARS-CoV2 nsp12 Rabbit Monoclonal Antibody [clone: EPR24837-213]
Anticorpos
Rabbit monoclonal [EPR24837-213] to SARS-CoV2 nsp12.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SARS-CoV2 nsp12
Clonality: Monoclonal
Clone: EPR24837-213
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR