Print…

Anti-SARS-CoV2 nsp12 Rabbit Monoclonal Antibody [clone: EPR24837-213]

Fornecedor: Abcam
AB315965-1MG AB315965-100UG
ABCAAB315965-1MGEA 9160 EUR
ABCAAB315965-1MG ABCAAB315965-100UG
Anti-SARS-CoV2 nsp12 Rabbit Monoclonal Antibody [clone: EPR24837-213]
Anticorpos
Rabbit monoclonal [EPR24837-213] to SARS-CoV2 nsp12 - BSA and Azide free.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: SARS-CoV2 nsp12
Clonality: Monoclonal
Clone: EPR24837-213
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR