Print…
Anti-YKL-40/CHI3L1 Rabbit Recombinant Multiclonal Antibody [clone: RM1204]
Fornecedor: Abcam
AB319164-20UL AB319164-100UL AB319164-1ML
ABCAAB319164-20ULEA 207 EUR
ABCAAB319164-20UL ABCAAB319164-100UL ABCAAB319164-1ML
Anti-YKL-40/CHI3L1 Rabbit Recombinant Multiclonal Antibody [clone: RM1204]
Anticorpos
Rabbit recombinant multiclonal [RM1204] to YKL-40/CHI3L1.
Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: YKL-40/CHI3L1
Clonality: Recombinant Multiclonal
Clone: RM1204
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat