Print…
Anti-Lactate Dehydrogenase A (LDHA) Rabbit Monoclonal Antibody [clone: EPR25915-88]
Fornecedor: Abcam
AB317641-100UG AB317641-1MG
ABCAAB317641-100UGEA 919 EUR
ABCAAB317641-100UG ABCAAB317641-1MG
Anti-Lactate Dehydrogenase A (LDHA) Rabbit Monoclonal Antibody [clone: EPR25915-88]
Anticorpos
Rabbit monoclonal [EPR25915-88] to Lactate Dehydrogenase A (LDHA) - BSA and Azide free (Detector).
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Lactate Dehydrogenase A (LDHA)
Clonality: Monoclonal
Clone: EPR25915-88
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human