Print…

Anti-Influenza A H1N1 (A/California/07/2009) neuraminidase Mouse Monoclonal Antibody [clone: CD6]

Fornecedor: Abcam
AB318988-100UG AB318988-20UG AB318988-1MG
ABCAAB318988-100UGEA 694 EUR
ABCAAB318988-100UG ABCAAB318988-20UG ABCAAB318988-1MG
Anti-Influenza A H1N1 (A/California/07/2009) neuraminidase Mouse Monoclonal Antibody [clone: CD6]
Anticorpos
Mouse monoclonal [CD6] to Influenza A H1N1 (A/California/07/2009) neuraminidase.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Influenza A H1N1 (A/California/07/2009) neuraminidase
Clonality: Monoclonal
Clone: CD6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR