Print…

Anti-PPP1R1A Rabbit Monoclonal Antibody [clone: EPR28192-90]

Fornecedor: Abcam
AB316931-20UL AB316931-100UL AB316931-1ML
ABCAAB316931-20ULEA 244 EUR
ABCAAB316931-20UL ABCAAB316931-100UL ABCAAB316931-1ML
Anti-PPP1R1A Rabbit Monoclonal Antibody [clone: EPR28192-90]
Anticorpos
Rabbit monoclonal [EPR28192-90] to PPP1R1A.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PPP1R1A
Clonality: Monoclonal
Clone: EPR28192-90
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR