Print…
Anti-SH2D1A/SAP Rat Monoclonal Antibody [clone: 1D12]
Fornecedor: Abcam
AB255862-1 AB255862-100
ABCAAB255862-1EA 9160 EUR
ABCAAB255862-1 ABCAAB255862-100
Anti-SH2D1A/SAP Rat Monoclonal Antibody [clone: 1D12]
Anticorpos
Rat monoclonal [1D12] to SH2D1A/SAP - BSA and Azide free.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: SH2D1A/SAP
Clonality: Monoclonal
Clone: 1D12
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human