Print…
Anti-AMH Rabbit Monoclonal Antibody [clone: EPR23380-171]
Fornecedor: Abcam
AB272221-100 AB272221-1ML
ABCAAB272221-100EA 788 EUR
ABCAAB272221-100 ABCAAB272221-1ML
Anti-AMH Rabbit Monoclonal Antibody [clone: EPR23380-171]
Anticorpos
Rabbit monoclonal [EPR23380-171] to AMH.
Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: AMH
Clonality: Monoclonal
Clone: EPR23380-171
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat