Print…

Anti-RELN Rabbit Monoclonal Antibody [clone: EPR23310-40]

Fornecedor: Abcam
AB281128-1 AB281128-100
ABCAAB281128-1EA 8680 EUR
ABCAAB281128-1 ABCAAB281128-100
Anti-RELN Rabbit Monoclonal Antibody [clone: EPR23310-40]
Anticorpos
Rabbit monoclonal [EPR23310-40] to Reelin - BSA and Azide free (Detector).

Type: Primary
Antigen: RELN
Clonality: Monoclonal
Clone: EPR23310-40
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR