Print…

Anti-Envelope Glycoprotein Rabbit Monoclonal Antibody [clone: 7H7]

Fornecedor: Abcam
AB288228-1 AB288228-100
ABCAAB288228-1EA 9160 EUR
ABCAAB288228-1 ABCAAB288228-100
Anti-Envelope Glycoprotein Rabbit Monoclonal Antibody [clone: 7H7]
Anticorpos
Rabbit monoclonal [7H7] to Envelope Glycoprotein - BSA and Azide free.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Envelope Glycoprotein
Clonality: Monoclonal
Clone: 7H7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zaire ebolavirus
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR