Print…

Anti-Cyclin D3/CCND3 Mouse Monoclonal Antibody [clone: 1/Cyclin D3]

Fornecedor: Abcam
AB289546-1MG AB289546-20UG
ABCAAB289546-1MGEA 6380 EUR
ABCAAB289546-1MG ABCAAB289546-20UG
Anti-Cyclin D3/CCND3 Mouse Monoclonal Antibody [clone: 1/Cyclin D3]
Anticorpos
Mouse monoclonal [1/Cyclin D3] to Cyclin D3/CCND3.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cyclin D3/CCND3
Clonality: Monoclonal
Clone: 1/Cyclin D3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Mouse;Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR