Print…

Anti-Histone H3 (tri methyl K79) Rabbit Monoclonal Antibody [clone: EPR17468(2)]

Fornecedor: Abcam
AB208189-100 AB208189-1ML
ABCAAB208189-100EA 625 EUR
ABCAAB208189-100 ABCAAB208189-1ML
Anti-Histone H3 (tri methyl K79) Rabbit Monoclonal Antibody [clone: EPR17468(2)]
Anticorpos
Rabbit monoclonal [EPR17468(2)] to Histone H3 (tri methyl K79) - ChIP Grade.

Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR17468(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR