Print…

Anti-ALK Rabbit Monoclonal Antibody [clone: EPR24750-116]

Fornecedor: Abcam
AB290733-100 AB290733-20UL AB290733-1ML
ABCAAB290733-100EA 707 EUR
ABCAAB290733-100 ABCAAB290733-20UL ABCAAB290733-1ML
Anti-ALK Rabbit Monoclonal Antibody [clone: EPR24750-116]
Anticorpos
Rabbit monoclonal [EPR24750-116] to ALK (phospho Y1604).

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ALK
Clonality: Monoclonal
Clone: EPR24750-116
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR