Print…

Anti-CD23 Rabbit Monoclonal Antibody [clone: 7] (FITC (Fluorescein Isothiocyanate))

Fornecedor: Abcam
AB275691-100 AB275691-25
ABCAAB275691-100EA 707 EUR
ABCAAB275691-100 ABCAAB275691-25
Anti-CD23 Rabbit Monoclonal Antibody [clone: 7] (FITC (Fluorescein Isothiocyanate))
Anticorpos
FITC Rabbit monoclonal [007] to CD23.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: 7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR