Print…

Anti-AMID Rat Monoclonal Antibody [clone: 6D8]

Fornecedor: Abcam
AB302673-10 AB302673-100 AB302673-20UG AB302673-1MG
ABCAAB302673-10EA 238 EUR
ABCAAB302673-10 ABCAAB302673-100 ABCAAB302673-20UG ABCAAB302673-1MG
Anti-AMID Rat Monoclonal Antibody [clone: 6D8]
Anticorpos
Rat monoclonal [6D8] to AMID.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: AMID
Clonality: Monoclonal
Clone: 6D8
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR