Print…

Anti-Nucleoside triphosphate phosphohydrolase Rabbit Monoclonal Antibody [clone: EPR14325]

Fornecedor: Abcam
AB182154-100 AB182154-1ML
ABCAAB182154-100EA 650 EUR
ABCAAB182154-100 ABCAAB182154-1ML
Anti-Nucleoside triphosphate phosphohydrolase Rabbit Monoclonal Antibody [clone: EPR14325]
Anticorpos
Rabbit monoclonal [EPR14325] to Nucleoside triphosphate phosphohydrolase.

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Nucleoside triphosphate phosphohydrolase
Clonality: Monoclonal
Clone: EPR14325
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR