Print…
Anti-HCF-1/Host Cell Factor C1 Rabbit Monoclonal Antibody [clone: EPR25290-18]
Fornecedor: Abcam
AB289975-100 AB289975-20UL AB289975-1ML
ABCAAB289975-100EA 707 EUR
ABCAAB289975-100 ABCAAB289975-20UL ABCAAB289975-1ML
Anti-HCF-1/Host Cell Factor C1 Rabbit Monoclonal Antibody [clone: EPR25290-18]
Anticorpos
Rabbit monoclonal [EPR25290-18] to HCF-1/Host Cell Factor C1.
Atenção: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: HCF-1 / Host Cell Factor C1
Clonality: Monoclonal
Clone: EPR25290-18
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat