Print…

Anti-Growth hormone receptor Rabbit Monoclonal Antibody [clone: EPR22481-266]

Fornecedor: Abcam
AB259485-1 AB259485-100
ABCAAB259485-1EA 9320 EUR
ABCAAB259485-1 ABCAAB259485-100
Anti-Growth hormone receptor Rabbit Monoclonal Antibody [clone: EPR22481-266]
Anticorpos
Rabbit monoclonal [EPR22481-266] to Growth hormone receptor - BSA and Azide free (Capture).

Atenção: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Growth hormone receptor
Clonality: Monoclonal
Clone: EPR22481-266
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR